Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrouridine

Supplier: BioVision
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-hydroxytetrahydropyrimidin-2(1H)-one

AB142080-25MG AB142080-10MG
10187-830EA 501.45 USD InStock
10187-830 10187-828
Tetrahydrouridine
Tetrahydrouridine


Formula: C₉H₁₆N₂O₆
MW: 248.2331 g/mol
MDL Number: MFCD00063468
CAS Number: 18771-50-1
Order Now
Specification Test Results
CAS 18771-50-1
Purity ~80% by TLC
Form Off-white solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty