Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thiazovivin

Supplier: BioVision
Synonyms: N-Benzyl-2-(pyrimidin-4-ylamino)thiazole-4-carboxamide

AB142081-5MG AB142081-1MG
10187-846EA 458.95 USD InStock
10187-846 10187-890
Thiazovivin
Thiazovivin


Formula: C₁₅H₁₃N₅OS
MW: 311,36 g/mol
MDL Number: MFCD16495823
CAS Number: 1226056-71-8
Order Now
Specification Test Results
CAS 1226056-71-8
Purity ≥98% by HPLC
Form Off-white solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty