Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capecitabine

Synonyms: 5'-Deoxy-5-fluoro-N4-[(pentyloxy)carbonyl]cytidinePentyl (1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl)carbamate

1741-1000 AB142097-100MG
10188-014EA 201.59 USD InStock
10188-014 10188-016
Capecitabine
Capecitabine


Formula: C₁₅H₂₂FN₃O₆
MW: 359.35 g/mol
MDL Number: MFCD00930626
CAS Number: 154361-50-9
Order Now
Specification Test Results
CAS 154361-50-9
Purity ≥99% by HPLC
Form White to off-white solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty