Would you like to log in again?
Puromycin dihydrochloride ≥98% (by HPLC)
Supplier: BioVision
AB141453-25MG 1860-500 AB141453-250MG AB141453-100MG 1860-1000
10188-328EA 811.42 USD InStock
10188-328 10191-152 10191-150 10188-330 10191-148
Puromycin dihydrochloride ≥98% (by HPLC)
Puromycin dihydrochloride
Puromycin Dihydrochloride is a nucleoside antibiotic
  • White to off-white powder
Puromycin has a fast mode of action, causing rapid cell death at low antibiotic concentrations. Adherent mammalian cells are sensitive to concentrations of 2 to 5 µg/mL, while cells in suspension are sensitive to concentrations as low as 0.5 to 2 µg/mL.
It inhibits protein synthesis by disrupting peptide transfer on ribosomes causing premature chain termination during translation. It is a potent translational inhibitor in both prokaryotic and eukaryotic cells.
Soluble in Water (50 mg/mL)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty