Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

StemRegenin 1 ≥98% (by HPLC)

Synonyms: SR14-(2-((2-(Benzo[b]thiophen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol4-(2-((2-Benzo[b]thiphen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol hydrochloride

AB142174-5MG AB142174-1MG
10191-214EA 689.5 USD InStock
10191-214 10191-212
StemRegenin 1 ≥98% (by HPLC)
StemRegenin 1
A cell-permeable purine derivative that acts as an antagonist of aryl hydrocarbon receptor and promotes the self-renewal of hematopoietic stem cells (HSCs). Culture of HSCs with SR1 led to a 50-fold increase in cells expressing CD34 and a 17-fold increase in cells that retain the ability to engraft immunodeficient mice. In the absence of cytokines, StemRegenin 1 did not induce proliferation, and at concentrations above 1 µM, it inhibits proliferation.

  • Appearance: White solid


Formula: C₂₄H₂₃N₅OS
MW: 429.542 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16987861
CAS Number: 1227633-49-9
Order Now
Specification Test Results
Purity ≥98% by HPLC
Form White solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty