Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

A77 1726

Synonyms: 2-Cyano-3-hydroxy-N-(4-(trifluoromethyl)phenyl)but-2-enamide

AB141480-25MG AB141480-5MG
10188-624EA 206.84 USD InStock
10188-624 10188-626
A77 1726
A77 1726


Formula: C₁₂H₉F₃N₂O₂
MW: 270.211 g/mol
MDL Number: MFCD00910058
CAS Number: 108605-62-5
Order Now
Specification Test Results
CAS 108605-62-5
Purity ≥98% by HPLC
Form White solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty