Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SANT-1

Supplier: BioVision
Synonyms: N-(4-Benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine

AB141483-5MG AB141483-1MG
10191-226EA 589.9 USD InStock
10191-226 10191-224
SANT-1
SANT-1


Formula: C₂₃H₂₇N₅
MW: 373.5 g/mol
MDL Number: MFCD01827306
CAS Number: 304909-07-7
Order Now
Specification Test Results
CAS 304909-07-7
Purity ≥98% by HPLC
Form Pale yellow solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty