Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperlonguminine

Synonyms: 5-(1,3-benzodioxol-5-yl)-n-(2-methylpropyl)penta-2,4-dienamide5-benzo[1,3]dioxol-5-yl-N- penta-2,4-dienamideN-Isobutylpiperamide

AB142185-50MG AB142185-10MG
10188-668EA 191.52 USD InStock
10188-668 10188-670
Piperlonguminine
Piperlonguminine


Formula: C₁₆H₁₉NO₃
MW: 273.332 g/mol
MDL Number: MFCD15146947
CAS Number: 5950-12-9
Order Now
Specification Test Results
Purity ≥98% by HPLC
Form White solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty