Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sunitinib free base

Supplier: BioVision
Synonyms: (Z)-N-(2-(Diethylamino)ethyl)-5-((5-fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide

AB285423-100MG AB285423-1G AB285423-25MG
10188-920EA 139.56 USD InStock
10188-920 10188-922 10188-924
Sunitinib free base
Sunitinib


Formula: C₂₂H₂₇FN₄O₂
MW: 398.47 g/mol
MDL Number: MFCD09260778
CAS Number: 557795-19-4
Order Now
Specification Test Results
CAS 557795-19-4
Purity ≥99% by HPLC
Form Yellow solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty