Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Torin 1

Supplier: BioVision
Synonyms: 1-(4-(4-Propionylpiperazin-1-yl)-3-(trifluoromethyl)phenyl)-9-(quinolin-3-yl)benzo[h][1,6]naphthyridin-2(1H)-one

AB218606-25MG AB218606-5MG
10189-392EA 252.88 USD InStock
10189-392 10189-394
Torin 1
Torin 1


Formula: C₃₅H₂₈F₃N₅O₂
MW: 607,62 g/mol
MDL Number: MFCD18782653
CAS Number: 1222998-36-8
Order Now
Specification Test Results
CAS 1222998-36-8
Purity ≥98% by HPLC
Form Slightly pinkish solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty