Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrouridine, EZSolution™

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-hydroxytetrahydropyrimidin-2(1H)-one

AB146617-25MG
10189-696EA 543.11 USD InStock
10189-696
Tetrahydrouridine, EZSolution™
Tetrahydrouridine


Formula: C₉H₁₆N₂O₆
MW: 248.2331 g/mol
MDL Number: MFCD00063468
CAS Number: 18771-50-1
Order Now
Specification Test Results
CAS 18771-50-1
Purity ~80% by TLC
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty