Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Blebbistatin

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


AB285433-5MG AB285433-1MG
10189-862EA 637.01 USD InStock
10189-862 10189-860
(+)-Blebbistatin
(+)-Blebbistatin


CAS Number: 1177356-70-5
Order Now
Specification Test Results
CAS 1177356-70-5
Purity ≥98% by HPLC
Form Yellow solid

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty