Would you like to log in again?
Human Recombinant Caspase-1
Supplier: BioVision
AB39901-25 AB39901-100
10190-140EA 489.14 USD InStock
10190-140 10190-142
Human Recombinant Caspase-1
Enzymes
Caspase-1 (also known as ICE) is a prototypical member of the caspase-family of cysteine proteases. Caspase-1 exists in cells as an inactive 45 kDa proenzyme. The pro-enzyme is matured by proteolysis to yield large (20 kD) and small (10 kD) subunits. The active caspase-1 is a heterotetramer consisting of two large and two small subunits. To date the regulatory mechanism of caspase-1 activation and the role of caspase-1 in apoptosis are poorly understood. In THP-1 cells, a large proportion of the caspase-1 is present in the inactive proenzyme form.
The recombinant active human caspase-1 was expressed in E. coli. The active caspase-1 preferentially cleaves caspase-1 substrates (e.g., YVAD-AFC or YVAD-pNA) and is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-YVAD-pNA (Cat. #10187-342) or Ac-YVAD-AFC (10187-338). The rh-Caspase has an N-term His-tag and corresponds to amino acids 120-404 of Caspase-1, gene accession no. NP_150634.1. The protein is provided as a minimum of 25 guaranteed units (Cat. No. 10190-142) or 100 guaranteed units (Cat. No. 10190-140).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty