Would you like to log in again?
Annexin V
Supplier: BioVision
AB14143-200 AB14143-1000
10190-890EA 1331.22 USD InStock
10190-890 10190-892
Annexin V
Proteins and Peptides
Annexin V/ANXA5-Cy3 Apoptosis Staining / Detection Reagent
Annexin V/ANXA5-Cy3 Apoptosis Detection Reagent can be used in a 10 min, one-step staining procedure to detect apoptosis by staining phosphatidylserine molecules which have translocated to the outside of the cell membrane. Analysis is by flow cytometry or fluorescence microscopy.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty