Would you like to log in again?
Annexin V (PE)
Supplier: BioVision
AB14154-200 AB14154-200
10190-910EA 452.49 USD InStock
10190-910 10190-908
Annexin V (PE)
Proteins and Peptides
Annexin V/ANXA5-PE Apoptosis Staining / Detection Reagent
Annexin V/ANXA5-PE Apoptosis Detection Reagent can be used in a 10 min, one-step staining procedure to detect apoptosis by staining phosphatidylserine molecules which have translocated to the outside of the cell membrane. Analysis is by flow cytometry or fluorescence microscopy.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty