Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 1 M in aqueous solution (pH 6.8)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Tris(hydroxymethyl)aminomethane hydrochlorideTris(hydroxymethyl)methylamine hydrochlorideTrizma(R) hydrochlorideTris(hydroxymethyl)methylammonium chlorideTHAM HClTrometamol hydrochloride2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochlorideTRIS HCl

AB286853-100ML
10190-924EA 180.43 USD InStock
10190-924
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 1 M in aqueous solution (pH 6.8)
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)


Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/01/2297601.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty