Would you like to log in again?
Orange G Loading Buffer, Biovision
Supplier: BioVision
AB286855-10ML
10190-932EA 158.36 USD InStock
10190-932
Orange G Loading Buffer, Biovision
Buffers Electrophoresis Buffers Loading Buffers
Orange G Dye runs faster than Bromophenol blue dye in standard agarose gel, which allows clear visualization of small DNA fragments on the gel.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty