Would you like to log in again?
IntraMedic™ Polyethylene Tubing, Stoelting
Supplier: Stoelting
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Intramedic™
51163 51154 51155 51150 51171 51151 51167
100229-432EA 294.23 USD InStock
100229-432 100229-422 95060-132 100228-596 100229-426 100229-434 100229-424
IntraMedic™ Polyethylene Tubing, Stoelting
Tubing Transfer Tubing
IntraMedic™ PE tubing is the lab standard in bioinert polyethylene tubing.
Use for attaching to syringe hubs for infusion, for catheterizing vessels, for tracheotomy, and for other general laboratory applications requiring small, strong, and durable tubing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty