Would you like to log in again?
Vernier Fan Cart
Supplier: VERNIER SOFTWARE & TECH SE
CART-FEC CART-F
470165-088EA 244.99 USD InStock
470165-088 470165-086
Vernier Fan Cart
Educational Materials Physics Educational Materials Mechanics Learning Activities
Collect quantitative acceleration data with this adjustable fan cart.
The fan has three speeds for studying the effect of variable thrust. Two mass bars let you change the total mass of the cart to observe how mass affects acceleration. The fan turns on a protractor base, allowing the thrust to be directed at a known angle from the direction of travel. In this way students can understand vector component applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty