Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SpinPrep™ Gel DNA Extraction Kit, MilliporeSigma
Supplier: MilliporeSigma
Millipore® SpinPrep™
70852-3 70852-3
EM70852-3EA 398.21 USD InStock
CA80500-380 EM70852-3
SpinPrep™ Gel DNA Extraction Kit, MilliporeSigma
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The SpinPrep™ Gel DNA Kit enables efficient extraction of DNA fragments of 150 bp to > 12,000 bp in size from agarose gels.
  • No organic extraction or alcohol precipitation
  • Total preparation time is less than 30 minutes
  • No low melting point agarose is required
The procedure uses GelMelt™ Solution to dissolve the gel slice, followed by adsorption of the DNA to a silica membrane in a spin column format. After a wash step, the purified DNA is eluted in low-salt buffer. Each spin column can bind up to 20 µg DNA. Recoveries are consistently in the range of 50-90% under standard conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty