Would you like to log in again?
Heart Rate Hand Grips
Supplier: VERNIER SOFTWARE & TECH SE
HR-GRIP
470165-124EA 40.99 USD InStock
470165-124
Heart Rate Hand Grips
Electrical Sockets
You can use our Heart Rate Hand Grips in conjunction with the Polar Transmitter Module. This option eliminates the need for a chest belt and allows students to measure their heart rate from the palm of their hands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty