Would you like to log in again?
Four Stroke Engine Model
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SA626
470163-996EA 1189 USD InStock
470163-996
Four Stroke Engine Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
The model is made up of full size four stroke, four cylinders, used in petrol engines.
  • Hand driven
  • Mounted on a sturdy acrylic frame
The model is manufactured using high quality raw material and ensure long service life and is corrosion resistant. The maximum parts like four cylinders of the cylinder block, cylinder heads, valve parts, crank shaft piston, cam, connecting rod inlet and exhaust manifolds, throttle body, spark plugs etc. are sanctioned to clearly demonstrate the internal constructional details.
The four stroke cycle are Intake, Compression, Power and Exhaust. Each corresponds to full stroke of the piston linked with revolutions of crank shaft.
Intake: The piston move downward drawing a fresh change of air and fuel vapor pressure mixture. Compression: The piston rises and propell valve is forced to shut by increased cylinder pressure. Power: At the top of compression stroke, the spark plug fires igniting the misture of air and fuel. Exhaust: At the bottom of power stroke, the exhaust valve is opened by the CAM/LITTER mechanism. The upward stroke of piston drive the exhausted fuel out of cylinder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty