Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-iodoethane ≥96%

Supplier: BeanTown Chemical

121110-5G 121110-25G 121110-1G
BT121110-5GEA 122.56 USD InStock
BT121110-5G BT121110-1G BT121110-25G
1-Chloro-2-iodoethane ≥96%
1-Chloro-2-iodoethane
  • Refractive index: 1.566
  • Light Sensitive
  • Liquid


Formula: C₂H₄ClI
MW: 190.41 g/mol
Density: 2.088 g/mL
MDL Number: MFCD00041453
CAS Number: 624-70-4
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty