Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) oxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Ferric oxideDiiron trioxideFerric Oxide, (iron(III) oxide)

121220-1KG 121220-250G 121220-50G
BT121220-50GEA 80.82 USD InStock
BT121220-50G BT121220-1KG BT121220-250G
Iron(III) oxide ≥99.9% (trace metals basis)
Iron (III) oxide
  • <5 Micron Powder


Formula: Fe₂O₃
MW: 159.69 g/mol
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty