Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5%

Supplier: BeanTown Chemical
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

121340-1KG 121340-5KG
BT121340-1KGEA 58.03 USD InStock
BT121340-1KG BT121340-5KG
Boric acid ≥99.5%
ortho-Boric acid
  • RTECS: ED4550000
  • Granular


Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: 160 °C (dec.)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty