Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Glutamic acid hydrochloride ≥99%

Supplier: BeanTown Chemical
Synonyms: (S)-2-Aminopentanedioic acid hydrochloride

122290-250G 122290-1KG
BT122290-1KGEA 67.31 USD InStock
BT122290-1KG BT122290-250G
L(+)-Glutamic acid hydrochloride ≥99%
L(+)-Glutamic acid hydrochloride
  • Hygroscopic
  • Crystalline


Formula: C₅H₁₀ClNO₄
MW: 183.59 g/mol
MDL Number: MFCD00012619
CAS Number: 138-15-8
EINECS: 205-315-9
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty