Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sesamol-(3,4-(methylenedioxy)phenol)

Supplier: BeanTown Chemical
Synonyms: Benzo[d][1,3]dioxol-5-olSesamol3,4-Methylenedioxyphenol

122420-250G 122420-50G 122420-10G
BT122420-10GEA 45.7 USD InStock
BT122420-10G BT122420-250G BT122420-50G
Sesamol-(3,4-(methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
  • RTECS: SM0890000
  • Crystalline


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 113…116 °C/2 mmHg
Melting Pt: 62…65 °C
MDL Number: MFCD00005827
CAS Number: 533-31-3
EINECS: 208-561-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty