Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexyl methyl ketone ≥95%

Supplier: BeanTown Chemical
Synonyms: 1-Cyclohexylethanone

122655-25G 122655-5G
BT122655-25GEA 126.73 USD InStock
BT122655-25G BT122655-5G
Cyclohexyl methyl ketone ≥95%
Cyclohexyl methyl ketone
  • RTECS: KM5638850
  • Liquid


Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 181…183 °C
Melting Pt: –34 °C
Density: 0.917 g/mL
Flash Pt: 66 °C (151 °F)
MDL Number: MFCD00040418
CAS Number: 823-76-7
EINECS: 212-517-0
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty