Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Threonine ≥99%

Supplier: BeanTown Chemical
Synonyms: (2R,3S)-2-Amino-3-hydroxybutanoic acidH-D-Thr-OH

122690-5G 122690-25G 122690-100G
BT122690-25GEA 100.76 USD InStock
BT122690-25G BT122690-5G BT122690-100G
D-Threonine ≥99%
D-Threonine
  • RTECS: XO8580000
  • Optical rotation: [α]20/D +29±1°, c = 5% in water
  • Crystalline


Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00064269
CAS Number: 632-20-2
EINECS: 211-171-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty