Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophthalic anhydride ≥95%

Supplier: BeanTown Chemical
Synonyms: 5-Nitroisobenzofuran-1,3-dione

126495-5G 126495-25G
BT126495-5GEA 38.31 USD InStock
BT126495-5G BT126495-25G
4-Nitrophthalic anhydride ≥95%
4-Nitrophthalic anhydride
  • RTECS: TI3328000
  • Moisture Sensitive
  • Powder


Formula: C₈H₃NO₅
MW: 193.12 g/mol
Melting Pt: 116…120 °C
MDL Number: MFCD00005922
CAS Number: 5466-84-2
EINECS: 226-776-2
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty