Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azelaic acid ≥98%

Supplier: BeanTown Chemical
Synonyms: 1,9-Nonanedioic acid1,7-Heptanedicarboxylic acid

144545-100G 144545-25G 144545-5X100G
BT144545-100GEA 79.43 USD InStock
BT144545-100G BT144545-25G BT144545-5X100
Azelaic acid ≥98%
Azelaic acid
  • RTECS: CM1980000
  • Crystalline


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 286 °C/100 mmHg
Melting Pt: 109…111 °C
Flash Pt: 210 °C (410 °F)
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty