Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Tagatose ≥98%

Supplier: BeanTown Chemical
Synonyms: (3S,4S,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one

128160-5G 128160-250MG 128160-1G
BT128160-1GEA 117.36 USD InStock
BT128160-1G BT128160-5G BT128160-250MG
D-Tagatose ≥98%
D-Tagatose
  • Powder


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00134449
CAS Number: 87-81-0
EINECS: 201-772-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty