Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium, gauze woven from wire 40 mesh Ø 0.127 mm

Supplier: BeanTown Chemical

129850-300X300 129850-75X75MM 129850-150X150
BT129850-75X75EA 224.33 USD InStock
BT129850-75X75 BT129850-300X3 BT129850-150X1
Titanium, gauze woven from wire 40 mesh Ø 0.127 mm
Titanium
  • Wire Cloth


Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty