Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver chloride, crystals optical disc, Length 25 mm, Width 4 mm

Synonyms: Silver(I) chloride

130510-1EACH
BT130510-1EACHEA 959.76 USD InStock
BT130510-1EACH
Silver chloride, crystals optical disc, Length 25 mm, Width 4 mm
Silver chloride
  • Optic Disc


Formula: AgCl
MW: 143.32 g/mol
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty