Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) hydroxide ≥94%, Technical Grade

Supplier: BeanTown Chemical

130790-2KG 130790-500G
BT130790-500GEA 72.77 USD InStock
BT130790-500G BT130790-2KG
Copper(II) hydroxide ≥94%, Technical Grade
Copper (II) hydroxide
  • RTECS: GL7600000
  • Hygroscopic
  • Powder


Formula: H₂CuO₂
MW: 97.56 g/mol
Density: 3.4 g/mL
MDL Number: MFCD00010968
CAS Number: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Assay 94%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty