Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neodymium(III) oxide ≥99% (trace rare earth metals basis)

Supplier: BeanTown Chemical
Synonyms: Dineodymium trioxideNeodymium oxide

130865-25G 130865-100G 130865-500G
BT130865-500GEA 493.38 USD InStock
BT130865-500G BT130865-25G BT130865-100G
Neodymium(III) oxide ≥99% (trace rare earth metals basis)
Neodymium (III) oxide
  • Hygroscopic
  • Powder


Formula: Nd₂O₃
MW: 336.48 g/mol
Melting Pt: 2272 °C
Density: 7.24 g/mL
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty