Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Serine ≥99%

Supplier: BeanTown Chemical
Synonyms: (±)-SerineH-DL-Ser-OH

131210-500G 131210-100G
BT131210-500GEA 190.07 USD InStock
BT131210-500G BT131210-100G
DL-Serine ≥99%
DL-Serine
  • Crystalline


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 240 °C (dec.)
MDL Number: MFCD00064223
CAS Number: 302-84-1
EINECS: 206-130-6
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty