Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 8,000

Supplier: BeanTown Chemical
Synonyms: PEG 8,000

132420-2KG 132420-500G 132420-100G
BT132420-2KGEA 141.01 USD InStock
BT132420-2KG BT132420-100G BT132420-500G
Polyethylene glycol 8,000
Polyethylene glycol 8,000
  • Powder


Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Melting Pt: 60…63 °C
MDL Number: MFCD01779615
CAS Number: 25322-68-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty