Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-amino-5-bromobenzoate ≥98%

Supplier: BeanTown Chemical
Synonyms: Methyl 2-amino-5-bromobenzoate

132815-5G 132815-100G 132815-25G
BT132815-5GEA 32.61 USD InStock
BT132815-5G BT132815-25G BT132815-100G
Methyl-2-amino-5-bromobenzoate ≥98%
Methyl 2-amino-5-bromobenzoate
  • Solid


Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Melting Pt: 72…74 °C
MDL Number: MFCD00144761
EINECS: 000-000-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty