Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Niobium(V) oxalate hydrate

Supplier: BeanTown Chemical

132875-500G 132875-25G 132875-100G
BT132875-500GEA 681.38 USD InStock
BT132875-500G BT132875-25G BT132875-100G
Niobium(V) oxalate hydrate
Niobium(V) oxalate hydrate
  • Powder


Formula: C₁₀H₅NbO₂₀·xH₂O
MW: 538.04 g/mol (anhy)
MDL Number: MFCD00050743
CAS Number: 21348-59-4
EINECS: 244-341-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty