Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3,5-dichlorobenzene ≥97%

Supplier: BeanTown Chemical
Synonyms: 3,5-Dichlorobromobenzene

133040-5G 133040-25G 133040-100G
BT133040-25GEA 64.81 USD InStock
BT133040-25G BT133040-100G BT133040-5G
1-Bromo-3,5-dichlorobenzene ≥97%
1-Bromo-3,5-dichlorobenzene
  • Powder


Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 232 °C
Melting Pt: 73…75 °C
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty