Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digallium trioxide ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

133185-25G 133185-100G 133185-5G
BT133185-100GEA 765.52 USD InStock
BT133185-100G BT133185-25G BT133185-5G
Digallium trioxide ≥99.999% (trace metals basis)
Digallium trioxide
  • -100 Mesh Powder


Formula: Ga₂O₃
MW: 187.44 g/mol
Melting Pt: 1900 °C
Density: 5.88 g/mL
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty