Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graphite ≥99.9995% (trace metals basis), rods, Length 30.5 cm, Diameter 6.15 mm.

Supplier: BeanTown Chemical

133350-25PC
BT133350-25PCEA 257.57 USD InStock
BT133350-25PC
Graphite ≥99.9995% (trace metals basis), rods, Length 30.5 cm, Diameter 6.15 mm.
Graphite
  • Solid, AGKSP grade, Ultra "F" purity


Formula: C
MW: 12.01 g/mol
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Order Now
Specification Test Results
Assay 99.9995%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty