Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Tolualdehyde ≥98% stabilized

Supplier: BeanTown Chemical
Synonyms: o-Tolualdehyde2-Methylbenzaldehyde

133365-100G 133365-25G 133365-500G
BT133365-500GEA 589.98 USD InStock
BT133365-500G BT133365-100G BT133365-25G
2-Tolualdehyde ≥98% stabilized
2-Tolualdehyde
  • Refractive index: 1.546
  • Air Sensitive
  • Liquid


Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 196…199 °C
Melting Pt: –35 °C
Density: 1.037 g/mL
Flash Pt: 67 °C (152 °F)
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty