Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Hydroxymethyl)benzyl alcohol ≥97%

Supplier: BeanTown Chemical
Synonyms: o-Phenylenedimethanol1,2-Benzenedimethanolo-Xylene-α,α'-diol

133425-1G 133425-5G 133425-25G
BT133425-5GEA 116.32 USD InStock
BT133425-5G BT133425-25G BT133425-1G
2-(Hydroxymethyl)benzyl alcohol ≥97%
2-(Hydroxymethyl)benzyl alcohol
  • Crystalline


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty