Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 1X8 (Cl) 100-200 (Cl), beads

Supplier: BeanTown Chemical

133815-250G 133815-1KG 133815-50G
BT133815-50GEA 40.7 USD InStock
BT133815-50G BT133815-1KG BT133815-250G
Dowex® 1X8 (Cl) 100-200 (Cl), beads
Dowex® 1X8 (Cl)


MDL Number: MFCD00132718
CAS Number: 12627-85-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty