Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triiodomethane ≥99%

Supplier: BeanTown Chemical
Synonyms: Iodoform

134040-250G 134040-1KG 134040-50G
BT134040-1KGEA 418.58 USD InStock
BT134040-1KG BT134040-250G BT134040-50G
Triiodomethane ≥99%
Triiodomethane
  • RTECS: PB7000000
  • Light Sensitive
  • Solid


Formula: CHI₃
MW: 393.73 g/mol
Melting Pt: 118…121 °C
Density: 4.008 g/mL
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty