Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin (II) sulfide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Tin sulphideTin sulfide

135005-25G 135005-5G
BT135005-25GEA 201.46 USD InStock
BT135005-25G BT135005-5G
Tin (II) sulfide ≥99.5% (trace metals basis)
Tin (II) sulphide
  • -4 Mesh Powder


Formula: SSn
MW: 150.78 g/mol
Melting Pt: 650 °C
Density: 5.22 g/mL
MDL Number: MFCD00011245
CAS Number: 1314-95-0
EINECS: 215-248-7
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty