Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Nonadecane ≥99%

Supplier: BeanTown Chemical
Synonyms: N-nonadecane

135135-25G 135135-100G
BT135135-25GEA 27.11 USD InStock
BT135135-25G BT135135-100G
n-Nonadecane ≥99%
n-Nonadecane
  • Refractive index: 1.441
  • Solid


Formula: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Boiling Pt: 440 °C
Melting Pt: 30…34 °C
Density: 0.786 g/mL
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty