Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Purine ≥98%

Supplier: BeanTown Chemical
Synonyms: 1H-Purine

135595-1G 135595-5G
BT135595-5GEA 674.11 USD InStock
BT135595-5G BT135595-1G
Purine ≥98%
Purine
  • RTECS: UO7450000
  • Crystalline/Powder


Formula: C₅H₄N₄
MW: 120.11 g/mol
Melting Pt: 214…217 °C
MDL Number: MFCD00079221
CAS Number: 120-73-0
EINECS: 204-421-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty